The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013848.3 | NP_001013870.1 | 18 - 19 | 1898 | 148 |
NM_001319194.1 | NP_001306123.1 | 18 - 19 | 1964 | 148 | |
NM_001319195.1 | NP_001306124.1 | 17 - 18 | 1849 | 148 | |
NM_001319200.1 | NP_001306129.1 | 13 - 14 | 1543 | 148 | |
NM_019053.5 | NP_061926.3 | 17 - 18 | 1852 | 148 | |
XM_017016344.1 | XP_016871833.1 | 18 - 19 | 1956 | 148 | |
XM_017016345.1 | XP_016871834.1 | 17 - 18 | 1933 | 148 | |
XM_017016346.1 | XP_016871835.1 | 17 - 18 | 1930 | 148 | |
XM_017016347.1 | XP_016871836.1 | 17 - 18 | 2136 | 148 | |
XM_017016348.1 | XP_016871837.1 | 18 - 19 | 1959 | 148 | |
GenBank mRNA | AK002113.1 | - | 9 - 10 | 850 | 148 |
AK123268.1 | - | 4 - 5 | 1207 | 148 | |
AK128190.1 | - | 18 - 19 | 1898 | 148 | |
AK293857.1 | - | 17 - 18 | 1815 | 148 | |
AK300285.1 | - | 18 - 19 | 1964 | 148 | |
AK315544.1 | - | 17 - 18 | 1787 | 148 | |
AL137438.1 | - | 12 - 13 | 1149 | 148 | |
AL832501.1 | - | 17 - 18 | 1793 | 148 | |
BC028395.2 | - | 17 - 18 | 1787 | 148 | |
EF571007.1 | - | 13 - 14 | 1484 | 148 |