The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021942.5 | NP_068761.4 | 16 - 17 | 1828 | 115 |
NM_199053.2 | NP_951008.1 | 16 - 17 | 1828 | 115 | |
XM_017008537.1 | XP_016864026.1 | 16 - 17 | 1828 | 115 | |
XM_017008538.1 | XP_016864027.1 | 16 - 17 | 1828 | 115 | |
XM_017008539.1 | XP_016864028.1 | 14 - 15 | 1479 | 115 | |
GenBank mRNA | AK091146.1 | - | 11 - 12 | 1052 | 115 |
AK096345.1 | - | 16 - 17 | 1802 | 115 | |
AK315057.1 | - | 11 - 12 | 1034 | 115 | |
AL136752.1 | - | 8 - 9 | 737 | 115 | |
AL833571.1 | - | 16 - 17 | 1807 | 115 | |
BC051724.1 | - | 16 - 17 | 1772 | 115 | |
BX538044.1 | - | 16 - 17 | 1782 | 115 | |
BX647127.1 | - | 16 - 17 | 1799 | 115 | |
BX648499.1 | - | 16 - 17 | 1803 | 115 | |
CR533553.1 | - | 8 - 9 | 672 | 115 |