The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166498.2 | NP_001159970.1 | 1 - 2 | 109 | 80 |
NM_006665.5 | NP_006656.2 | 1 - 2 | 109 | 80 | |
GenBank mRNA | AF155510.1 | - | 1 - 2 | 108 | 80 |
BC051321.1 | - | 1 - 2 | 109 | 80 | |
FJ517659.1 | - | 1 - 2 | 109 | 80 | |
FJ517660.1 | - | 1 - 2 | 109 | 80 | |
GQ337901.1 | - | 1 - 2 | 109 | 80 | |
GQ337902.1 | - | 1 - 2 | 109 | 80 | |
HM187643.1 | - | 1 - 2 | 109 | 80 |