The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198721.3 | NP_942014.1 | 35 - 36 | 2377 | 76 |
XM_017008735.1 | XP_016864224.1 | 35 - 36 | 2238 | 76 | |
XM_017008736.1 | XP_016864225.1 | 34 - 35 | 2573 | 76 | |
GenBank mRNA | AF293340.1 | - | 35 - 36 | 2377 | 76 |