The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153646.3 | NP_705932.2 | 1 - 2 | 152 | 55 |
NM_153647.3 | NP_705933.2 | 2 - 3 | 225 | 55 | |
GenBank mRNA | AF520704.1 | - | 1 - 2 | 102 | 55 |
AF520705.1 | - | 1 - 2 | 102 | 55 | |
AK295059.1 | - | 2 - 3 | 225 | 55 | |
BC069653.1 | - | 1 - 2 | 102 | 55 |