The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135648.2 | NP_001129120.1 | 22 - 23 | 3428 | 104 |
NM_001291981.1 | NP_001278910.1 | 24 - 25 | 3476 | 104 | |
NM_001291984.1 | NP_001278913.1 | 21 - 22 | 3407 | 104 | |
NM_002844.3 | NP_002835.2 | 21 - 22 | 3410 | 104 | |
XM_006715537.2 | XP_006715600.1 | 22 - 23 | 3476 | 104 | |
XM_011536014.2 | XP_011534316.1 | 24 - 25 | 3524 | 104 | |
XM_011536015.2 | XP_011534317.1 | 23 - 24 | 3515 | 104 | |
XM_011536016.2 | XP_011534318.1 | 23 - 24 | 3151 | 104 | |
XM_011536017.2 | XP_011534319.1 | 25 - 26 | 3767 | 104 | |
XM_011536018.2 | XP_011534320.1 | 25 - 26 | 3440 | 104 | |
XM_017011145.1 | XP_016866634.1 | 23 - 24 | 3443 | 104 | |
XM_017011146.1 | XP_016866635.1 | 23 - 24 | 3392 | 104 | |
XM_017011147.1 | XP_016866636.1 | 23 - 24 | 3370 | 104 | |
XM_017011148.1 | XP_016866637.1 | 22 - 23 | 3349 | 104 | |
XM_017011149.1 | XP_016866638.1 | 12 - 13 | 1028 | 104 | |
XM_017011150.1 | XP_016866639.1 | 11 - 12 | 1015 | 104 | |
GenBank mRNA | AF533875.1 | - | 21 - 22 | 3175 | 104 |
BC140775.1 | - | 22 - 23 | 3230 | 104 | |
BC144512.1 | - | 22 - 23 | 3230 | 104 | |
BC144513.1 | - | 24 - 25 | 3278 | 104 | |
CR749277.1 | - | 21 - 22 | 3351 | 104 | |
L77886.1 | - | 21 - 22 | 3109 | 104 | |
Z70660.1 | - | 21 - 22 | 3091 | 104 |