Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PTPRK
- Entrez Gene ID
-
5796
- Gene Name
- protein tyrosine phosphatase, receptor type K
- Gene Aliases
-
R-PTP-kappa
- Chromosome Location
-
Chr.6: 127968779 - 128520847 on Build GRCh38
View Full Product Details