The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135648.2 | NP_001129120.1 | 1 - 2 | 415 | 91 |
NM_001291981.1 | NP_001278910.1 | 1 - 2 | 415 | 91 | |
NM_001291982.1 | NP_001278911.1 | 1 - 2 | 415 | 91 | |
NM_001291983.1 | NP_001278912.1 | 1 - 2 | 415 | 91 | |
NM_001291984.1 | NP_001278913.1 | 1 - 2 | 415 | 91 | |
NM_002844.3 | NP_002835.2 | 1 - 2 | 415 | 91 | |
XM_006715537.2 | XP_006715600.1 | 1 - 2 | 466 | 91 | |
XM_011536014.2 | XP_011534316.1 | 1 - 2 | 466 | 91 | |
XM_011536015.2 | XP_011534317.1 | 1 - 2 | 466 | 91 | |
XM_011536016.2 | XP_011534318.1 | 1 - 2 | 108 | 91 | |
XM_011536020.2 | XP_011534322.1 | 1 - 2 | 466 | 91 | |
XM_011536021.2 | XP_011534323.1 | 1 - 2 | 466 | 91 | |
GenBank mRNA | AF533875.1 | - | 1 - 2 | 180 | 91 |
AK295028.1 | - | 1 - 2 | 340 | 91 | |
BC140775.1 | - | 1 - 2 | 217 | 91 | |
BC144512.1 | - | 1 - 2 | 217 | 91 | |
BC144513.1 | - | 1 - 2 | 217 | 91 | |
CR749277.1 | - | 1 - 2 | 356 | 91 | |
DB020920.1 | - | 1 - 2 | 416 | 91 | |
L77886.1 | - | 1 - 2 | 114 | 91 | |
Z70660.1 | - | 1 - 2 | 99 | 91 |