The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016021.2 | NP_057105.2 | 1 - 2 | 362 | 60 |
XM_011535887.1 | XP_011534189.1 | 1 - 2 | 349 | 60 | |
XM_011535888.2 | XP_011534190.1 | 1 - 2 | 379 | 60 | |
GenBank mRNA | AF151834.1 | - | 1 - 2 | 169 | 60 |
AF161502.1 | - | 1 - 2 | 57 | 60 | |
AJ245898.1 | - | 1 - 2 | 93 | 60 | |
AK223464.1 | - | 2 - 3 | 101 | 60 | |
AK290574.1 | - | 1 - 2 | 204 | 60 | |
BC013973.1 | - | 1 - 2 | 324 | 60 | |
BF058850.1 | - | 2 - 3 | 281 | 60 | |
U93243.1 | - | 1 - 2 | 58 | 60 |