The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017525.2 | NP_059995.2 | 33 - 34 | 4271 | 57 |
XM_011545155.2 | XP_011543457.1 | 33 - 34 | 4440 | 57 | |
XM_011545156.2 | XP_011543458.1 | 33 - 34 | 4420 | 57 | |
XM_011545157.2 | XP_011543459.1 | 32 - 33 | 4343 | 57 | |
XM_011545158.2 | XP_011543460.1 | 32 - 33 | 4343 | 57 | |
XM_011545159.2 | XP_011543461.1 | 33 - 34 | 4441 | 57 | |
XM_011545160.2 | XP_011543462.1 | 34 - 35 | 4392 | 57 | |
XM_017017996.1 | XP_016873485.1 | 33 - 34 | 4437 | 57 | |
XM_017017999.1 | XP_016873488.1 | 23 - 24 | 3020 | 57 | |
GenBank mRNA | AY648038.1 | - | 33 - 34 | 4271 | 57 |