The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017525.2 | NP_059995.2 | 3 - 4 | 343 | 73 |
XM_011545155.2 | XP_011543457.1 | 3 - 4 | 458 | 73 | |
XM_011545156.2 | XP_011543458.1 | 3 - 4 | 459 | 73 | |
XM_011545157.2 | XP_011543459.1 | 3 - 4 | 459 | 73 | |
XM_011545158.2 | XP_011543460.1 | 3 - 4 | 459 | 73 | |
XM_011545159.2 | XP_011543461.1 | 3 - 4 | 459 | 73 | |
XM_011545160.2 | XP_011543462.1 | 4 - 5 | 410 | 73 | |
XM_011545161.2 | XP_011543463.1 | 3 - 4 | 459 | 73 | |
XM_017017996.1 | XP_016873485.1 | 3 - 4 | 458 | 73 | |
XM_017017997.1 | XP_016873486.1 | 3 - 4 | 459 | 73 | |
XM_017017998.1 | XP_016873487.1 | 3 - 4 | 459 | 73 | |
GenBank mRNA | AY648038.1 | - | 3 - 4 | 343 | 73 |