The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170794.1 | NP_001164265.1 | 1 - 2 | 433 | 91 |
XM_017011162.1 | XP_016866651.1 | 1 - 2 | 437 | 91 | |
GenBank mRNA | AB208889.1 | - | 1 - 2 | 70 | 91 |
DB077323.1 | - | 1 - 2 | 368 | 91 |