The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002553.3 | NP_002544.1 | 9 - 10 | 1021 | 120 |
GenBank mRNA | AF047599.1 | - | 9 - 10 | 956 | 120 |
AF049127.1 | - | 9 - 10 | 952 | 120 | |
AK223361.1 | - | 9 - 10 | 1021 | 120 | |
AK292285.1 | - | 9 - 10 | 997 | 120 | |
AK302122.1 | - | 10 - 11 | 1108 | 120 | |
AK313596.1 | - | 9 - 10 | 978 | 120 | |
BC023652.2 | - | 9 - 10 | 959 | 120 | |
U92538.1 | - | 9 - 10 | 966 | 120 |