The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127217.2 | NP_001120689.1 | 4 - 5 | 1121 | 131 |
XM_005266403.3 | XP_005266460.1 | 3 - 4 | 824 | 131 | |
XM_006719827.3 | XP_006719890.1 | 4 - 5 | 1051 | 131 | |
GenBank mRNA | AK311686.1 | - | 2 - 3 | 156 | 131 |
AK314920.1 | - | 3 - 4 | 830 | 131 | |
D83760.1 | - | 3 - 4 | 833 | 131 |