Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BACH2
Entrez Gene ID
60468
Gene Name
BTB domain and CNC homolog 2
Gene Aliases
BTBD25
Chromosome Location
Chr.6: 89926528 - 90296912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021813.3 NP_068585.1 2 - 3 520 76
XM_017011156.1 XP_016866645.1 2 - 3 524 76
XM_017011157.1 XP_016866646.1 2 - 3 524 76
XM_017011158.1 XP_016866647.1 2 - 3 524 76
XM_017011159.1 XP_016866648.1 2 - 3 524 76
XM_017011161.1 XP_016866650.1 2 - 3 524 76
XM_017011166.1 XP_016866655.1 2 - 3 524 76
XM_017011167.1 XP_016866656.1 2 - 3 524 76
GenBank mRNA AA885077.1 - 2 - 3 350 76
AI524722.1 - 2 - 3 354 76
AJ271878.1 - 2 - 3 354 76

View Full Product Details