The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031291.3 | NP_112581.1 | 5 - 6 | 924 | 86 |
XM_011532298.1 | XP_011530600.1 | 3 - 4 | 654 | 86 | |
GenBank mRNA | AJ863129.1 | - | 5 - 6 | 756 | 86 |
AL136857.1 | - | 5 - 6 | 883 | 86 | |
AY550240.1 | - | 5 - 6 | 875 | 86 | |
BC022032.2 | - | 5 - 6 | 875 | 86 | |
BU853653.1 | - | 2 - 3 | 161 | 86 |