Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLRB1
Entrez Gene ID
3820
Gene Name
killer cell lectin like receptor B1
Gene Aliases
CD161, CLEC5B, NKR, NKR-P1, NKR-P1A, NKRP1A, hNKR-P1A
Chromosome Location
Chr.12: 9595274 - 9607901 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002258.2 NP_002249.1 2 - 3 249 90
GenBank mRNA AK292022.1 - 2 - 3 264 90
BC113997.1 - 2 - 3 237 90
BC114516.1 - 2 - 3 237 90
BF432813.1 - 2 - 3 250 90
U11276.1 - 2 - 3 247 90

View Full Product Details