The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002432.1 | NP_002423.1 | 5 - 6 | 1193 | 112 |
GenBank mRNA | AK290392.1 | - | 5 - 6 | 1200 | 112 |
AK313210.1 | - | 5 - 6 | 1204 | 112 | |
BC032319.1 | - | 5 - 6 | 1204 | 112 | |
M81750.1 | - | 5 - 6 | 1193 | 112 |