Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC5
Entrez Gene ID
25921
Gene Name
zinc finger DHHC-type containing 5
Gene Aliases
DHHC5, ZNF375
Chromosome Location
Chr.11: 57667836 - 57701187 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015457.2 NP_056272.2 2 - 3 1362 63
XM_011544899.1 XP_011543201.1 2 - 3 1292 63
XM_011544900.1 XP_011543202.1 2 - 3 1200 63
XM_011544901.1 XP_011543203.1 2 - 3 1323 63
GenBank mRNA AB051535.1 - 2 - 3 1354 63
AK023130.1 - 1 - 2 519 63
AY894889.1 - 2 - 3 1468 63

View Full Product Details