The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003668.3 | NP_003659.2 | 5 - 6 | 896 | 65 |
NM_139078.2 | NP_620777.1 | 5 - 6 | 896 | 65 | |
XM_017020125.1 | XP_016875614.1 | 5 - 6 | 388 | 65 | |
XM_017020127.1 | XP_016875616.1 | 4 - 5 | 721 | 65 | |
XM_017020128.1 | XP_016875617.1 | 4 - 5 | 721 | 65 | |
XM_017020129.1 | XP_016875618.1 | 4 - 5 | 558 | 65 | |
XM_017020130.1 | XP_016875619.1 | 4 - 5 | 326 | 65 | |
XM_017020131.1 | XP_016875620.1 | 4 - 5 | 721 | 65 | |
GenBank mRNA | AK122767.1 | - | 5 - 6 | 1146 | 65 |
BC000833.1 | - | 4 - 5 | 339 | 65 | |
BC041049.1 | - | 5 - 6 | 883 | 65 | |
BC047284.1 | - | 5 - 6 | 564 | 65 |