The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142523.1 | NP_001135995.1 | 4 - 5 | 503 | 85 |
NM_007199.2 | NP_009130.2 | 5 - 6 | 686 | 85 | |
GenBank mRNA | AF113136.1 | - | 5 - 6 | 647 | 85 |
AK298645.1 | - | 4 - 5 | 444 | 85 | |
AK314040.1 | - | 5 - 6 | 668 | 85 | |
BC057800.1 | - | 5 - 6 | 661 | 85 | |
BC069388.1 | - | 5 - 6 | 634 | 85 |