Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIGLEC5
Entrez Gene ID
8778
Gene Name
sialic acid binding Ig like lectin 5
Gene Aliases
CD170, CD33L2, OB-BP2, OBBP2, SIGLEC-5
Chromosome Location
Chr.19: 51610967 - 51630474 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003830.3 NP_003821.1 5 - 6 1142 135
XM_011527438.2 XP_011525740.1 5 - 6 1096 135
XM_017027419.1 XP_016882908.1 5 - 6 1096 135
GenBank mRNA AK225753.1 - 5 - 6 1063 135
BC029896.1 - 5 - 6 1044 135
U71383.1 - 5 - 6 1145 135

View Full Product Details