Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIGLEC5
Entrez Gene ID
8778
Gene Name
sialic acid binding Ig like lectin 5
Gene Aliases
CD170, CD33L2, OB-BP2, OBBP2, SIGLEC-5
Chromosome Location
Chr.19: 51610967 - 51630474 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003830.3 NP_003821.1 6 - 7 1421 96
XM_011527438.2 XP_011525740.1 6 - 7 1375 96
XM_017027419.1 XP_016882908.1 6 - 7 1375 96
GenBank mRNA AF170484.1 - 6 - 7 1318 96
AK225753.1 - 6 - 7 1342 96
BC029896.1 - 6 - 7 1323 96
U71383.1 - 6 - 7 1424 96

View Full Product Details