The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001959.1 | NP_001001959.1 | 1 - 1 | 735 | 74 |
GenBank mRNA | BC140753.1 | - | 1 - 1 | 791 | 74 |