The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161440.2 | NP_001154912.2 | 12 - 13 | 2104 | 73 |
NM_002842.4 | NP_002833.4 | 14 - 15 | 2638 | 73 | |
XM_011527183.1 | XP_011525485.1 | 14 - 15 | 2659 | 73 | |
XM_011527188.1 | XP_011525490.1 | 13 - 14 | 2608 | 73 | |
XM_017027056.1 | XP_016882545.1 | 13 - 14 | 2646 | 73 | |
XM_017027057.1 | XP_016882546.1 | 13 - 14 | 2634 | 73 | |
XM_017027058.1 | XP_016882547.1 | 13 - 14 | 2679 | 73 | |
XM_017027059.1 | XP_016882548.1 | 13 - 14 | 2646 | 73 | |
XM_017027060.1 | XP_016882549.1 | 12 - 13 | 2598 | 73 | |
XM_017027061.1 | XP_016882550.1 | 11 - 12 | 2112 | 73 | |
XM_017027062.1 | XP_016882551.1 | 13 - 14 | 2554 | 73 | |
XM_017027063.1 | XP_016882552.1 | 13 - 14 | 2547 | 73 | |
GenBank mRNA | BC111715.1 | - | 13 - 14 | 2054 | 73 |
BC111716.1 | - | 14 - 15 | 2588 | 73 | |
D15049.2 | - | 14 - 15 | 2612 | 73 |