The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161440.2 | NP_001154912.2 | 7 - 8 | 1531 | 84 |
NM_002842.4 | NP_002833.4 | 9 - 10 | 2065 | 84 | |
XM_011527183.1 | XP_011525485.1 | 9 - 10 | 2086 | 84 | |
XM_011527188.1 | XP_011525490.1 | 8 - 9 | 2035 | 84 | |
XM_011527190.1 | XP_011525492.1 | 9 - 10 | 2086 | 84 | |
XM_017027056.1 | XP_016882545.1 | 8 - 9 | 2073 | 84 | |
XM_017027058.1 | XP_016882547.1 | 8 - 9 | 2106 | 84 | |
XM_017027059.1 | XP_016882548.1 | 8 - 9 | 2073 | 84 | |
XM_017027060.1 | XP_016882549.1 | 8 - 9 | 2073 | 84 | |
XM_017027061.1 | XP_016882550.1 | 6 - 7 | 1539 | 84 | |
XM_017027062.1 | XP_016882551.1 | 8 - 9 | 1981 | 84 | |
XM_017027063.1 | XP_016882552.1 | 8 - 9 | 1974 | 84 | |
XM_017027064.1 | XP_016882553.1 | 8 - 9 | 2073 | 84 | |
GenBank mRNA | AK097257.1 | - | 9 - 10 | 2084 | 84 |
BC111715.1 | - | 8 - 9 | 1481 | 84 | |
BC111716.1 | - | 9 - 10 | 2015 | 84 | |
D15049.2 | - | 9 - 10 | 2039 | 84 |