The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170931.1 | NP_001164402.1 | 3 - 4 | 1725 | 55 |
NM_005938.3 | NP_005929.2 | 2 - 3 | 1890 | 55 | |
XM_011530957.2 | XP_011529259.1 | 2 - 3 | 1307 | 55 | |
GenBank mRNA | BC026735.1 | - | 1 - 2 | 999 | 55 |
BC106761.2 | - | 2 - 3 | 1616 | 55 | |
X93996.1 | - | 2 - 3 | 1680 | 55 |