Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL23A1
Entrez Gene ID
91522
Gene Name
collagen type XXIII alpha 1 chain
Gene Aliases
-
Chromosome Location
Chr.5: 178237616 - 178590555 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173465.3 NP_775736.2 9 - 10 997 84
XM_006714933.3 XP_006714996.1 9 - 10 727 84
XM_006714934.3 XP_006714997.1 8 - 9 700 84
XM_006714936.3 XP_006714999.1 9 - 10 727 84
XM_011534688.2 XP_011532990.1 8 - 9 700 84
XM_011534689.2 XP_011532991.1 9 - 10 727 84
XM_011534690.2 XP_011532992.1 7 - 8 673 84
XM_011534691.2 XP_011532993.1 5 - 6 619 84
XM_011534692.2 XP_011532994.1 9 - 10 1752 84
XM_017010018.1 XP_016865507.1 9 - 10 727 84
GenBank mRNA AY158895.1 - 9 - 10 997 84
AY898961.1 - 9 - 10 997 84
BC042428.1 - 9 - 10 676 84

View Full Product Details