The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291978.1 | NP_001278907.1 | 2 - 3 | 517 | 125 |
NM_001291979.1 | NP_001278908.1 | 2 - 3 | 517 | 125 | |
NM_003703.2 | NP_003694.1 | 2 - 3 | 517 | 125 | |
GenBank mRNA | AF040965.1 | - | 2 - 3 | 379 | 125 |
AK292939.1 | - | 2 - 3 | 517 | 125 | |
BC026035.1 | - | 2 - 3 | 344 | 125 | |
BC053635.1 | - | 2 - 3 | 396 | 125 |