Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESRP1
Entrez Gene ID
54845
Gene Name
epithelial splicing regulatory protein 1
Gene Aliases
RBM35A, RMB35A
Chromosome Location
Chr.8: 94640999 - 94707466 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034915.2 NP_001030087.2 2 - 3 445 83
NM_001122825.1 NP_001116297.1 2 - 3 445 83
NM_001122826.1 NP_001116298.1 2 - 3 445 83
NM_001122827.1 NP_001116299.1 2 - 3 445 83
NM_017697.3 NP_060167.2 2 - 3 445 83
XM_005250991.3 XP_005251048.1 2 - 3 582 83
XM_005250992.3 XP_005251049.1 2 - 3 582 83
GenBank mRNA BC019932.2 - 2 - 3 406 83
BC067098.1 - 2 - 3 399 83
BC099916.1 - 2 - 3 409 83

View Full Product Details