The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034915.2 | NP_001030087.2 | 9 - 10 | 1113 | 69 |
NM_001122825.1 | NP_001116297.1 | 9 - 10 | 1113 | 69 | |
NM_001122826.1 | NP_001116298.1 | 9 - 10 | 1113 | 69 | |
NM_001122827.1 | NP_001116299.1 | 9 - 10 | 1113 | 69 | |
NM_017697.3 | NP_060167.2 | 9 - 10 | 1113 | 69 | |
XM_005250991.3 | XP_005251048.1 | 9 - 10 | 1250 | 69 | |
XM_005250992.3 | XP_005251049.1 | 9 - 10 | 1250 | 69 | |
GenBank mRNA | AK000178.1 | - | 6 - 7 | 507 | 69 |
BC019932.2 | - | 9 - 10 | 1074 | 69 | |
BC035406.1 | - | 6 - 7 | 526 | 69 | |
BC067098.1 | - | 9 - 10 | 1067 | 69 | |
BC099916.1 | - | 9 - 10 | 1077 | 69 | |
BC101621.1 | - | 6 - 7 | 472 | 69 | |
BC112043.1 | - | 6 - 7 | 472 | 69 | |
BX647570.1 | - | 9 - 10 | 1175 | 69 |