Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST5
Entrez Gene ID
6764
Gene Name
suppression of tumorigenicity 5
Gene Aliases
DENND2B, HTS1, p126
Chromosome Location
Chr.11: 8693352 - 8910951 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005418.3 NP_005409.3 17 - 18 3109 100
NM_139157.2 NP_631896.1 13 - 14 1577 100
NM_213618.1 NP_998783.1 14 - 15 2909 100
XM_005253077.1 XP_005253134.1 14 - 15 2824 100
XM_005253083.2 XP_005253140.1 13 - 14 1719 100
XM_011520309.1 XP_011518611.1 15 - 16 2884 100
XM_011520310.1 XP_011518612.1 15 - 16 3296 100
XM_011520311.2 XP_011518613.1 15 - 16 3278 100
XM_011520312.1 XP_011518614.1 19 - 20 3243 100
XM_011520313.1 XP_011518615.1 18 - 19 3129 100
XM_011520314.1 XP_011518616.1 17 - 18 3021 100
XM_011520315.1 XP_011518617.1 17 - 18 3117 100
XM_011520316.1 XP_011518618.1 16 - 17 3047 100
XM_011520317.1 XP_011518619.1 15 - 16 2988 100
XM_011520318.1 XP_011518620.1 18 - 19 3183 100
XM_011520319.2 XP_011518621.1 15 - 16 2836 100
XM_011520323.2 XP_011518625.2 14 - 15 1707 100
XM_011520324.2 XP_011518626.2 13 - 14 1647 100
XM_011520325.1 XP_011518627.1 14 - 15 1779 100
XM_011520326.1 XP_011518628.1 18 - 19 1983 100
XM_011520327.1 XP_011518629.1 14 - 15 1672 100
XM_011520328.1 XP_011518630.1 14 - 15 1704 100
XM_011520329.1 XP_011518631.1 17 - 18 1923 100
XM_017018182.1 XP_016873671.1 16 - 17 3002 100
XM_017018183.1 XP_016873672.1 16 - 17 2979 100
XM_017018184.1 XP_016873673.1 18 - 19 3138 100
XM_017018185.1 XP_016873674.1 14 - 15 2934 100
XM_017018186.1 XP_016873675.1 13 - 14 2874 100
XM_017018187.1 XP_016873676.1 14 - 15 1643 100
GenBank mRNA AK127763.1 - 11 - 12 1964 100
AK293245.1 - 11 - 12 1342 100
AK307617.1 - 3 - 4 464 100
AK312305.1 - 14 - 15 2875 100
AK312758.1 - 13 - 14 1919 100
BC036655.1 - 17 - 18 3114 100
BX647431.1 - 14 - 15 2909 100
S45936.1 - 10 - 11 1247 100
U15131.1 - 14 - 15 2837 100
U15779.1 - 10 - 11 1206 100
U15780.1 - 13 - 14 1577 100

View Full Product Details