Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST5
Entrez Gene ID
6764
Gene Name
suppression of tumorigenicity 5
Gene Aliases
DENND2B, HTS1, p126
Chromosome Location
Chr.11: 8693352 - 8910951 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005418.3 NP_005409.3 22 - 23 3767 102
NM_139157.2 NP_631896.1 18 - 19 2235 102
NM_213618.1 NP_998783.1 19 - 20 3567 102
XM_005253077.1 XP_005253134.1 19 - 20 3482 102
XM_005253083.2 XP_005253140.1 18 - 19 2377 102
XM_011520309.1 XP_011518611.1 20 - 21 3542 102
XM_011520310.1 XP_011518612.1 20 - 21 3954 102
XM_011520311.2 XP_011518613.1 20 - 21 3936 102
XM_011520312.1 XP_011518614.1 24 - 25 3901 102
XM_011520313.1 XP_011518615.1 23 - 24 3787 102
XM_011520314.1 XP_011518616.1 22 - 23 3679 102
XM_011520315.1 XP_011518617.1 22 - 23 3775 102
XM_011520316.1 XP_011518618.1 21 - 22 3705 102
XM_011520317.1 XP_011518619.1 20 - 21 3646 102
XM_011520318.1 XP_011518620.1 23 - 24 3841 102
XM_011520319.2 XP_011518621.1 20 - 21 3494 102
XM_011520323.2 XP_011518625.2 19 - 20 2365 102
XM_011520324.2 XP_011518626.2 18 - 19 2305 102
XM_011520325.1 XP_011518627.1 19 - 20 2437 102
XM_011520326.1 XP_011518628.1 23 - 24 2641 102
XM_011520327.1 XP_011518629.1 19 - 20 2330 102
XM_011520328.1 XP_011518630.1 19 - 20 2362 102
XM_011520329.1 XP_011518631.1 22 - 23 2581 102
XM_017018182.1 XP_016873671.1 21 - 22 3660 102
XM_017018183.1 XP_016873672.1 21 - 22 3637 102
XM_017018184.1 XP_016873673.1 23 - 24 3796 102
XM_017018185.1 XP_016873674.1 19 - 20 3592 102
XM_017018186.1 XP_016873675.1 18 - 19 3532 102
XM_017018187.1 XP_016873676.1 19 - 20 2301 102
GenBank mRNA AK127763.1 - 16 - 17 2622 102
AK293245.1 - 16 - 17 2000 102
AK307617.1 - 8 - 9 1122 102
BC036655.1 - 22 - 23 3772 102
BX647431.1 - 19 - 20 3567 102
S45936.1 - 15 - 16 1905 102
U15131.1 - 19 - 20 3495 102
U15779.1 - 15 - 16 1864 102
U15780.1 - 18 - 19 2235 102

View Full Product Details