The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142548.1 | NP_001136020.1 | 3 - 4 | 203 | 91 |
NM_003579.3 | NP_003570.2 | 2 - 3 | 800 | 91 | |
XM_006710975.3 | XP_006711038.1 | 2 - 3 | 138 | 91 | |
GenBank mRNA | AK292611.1 | - | 3 - 4 | 183 | 91 |
BC121059.1 | - | 2 - 3 | 308 | 91 | |
BC121060.1 | - | 2 - 3 | 308 | 91 | |
X97795.1 | - | 2 - 3 | 189 | 91 |