The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005739.1 | NP_001005739.1 | 19 - 20 | 2672 | 160 |
NM_016516.2 | NP_057600.2 | 19 - 20 | 2708 | 160 | |
XM_006712029.3 | XP_006712092.1 | 18 - 19 | 2568 | 160 | |
XM_017004316.1 | XP_016859805.1 | 18 - 19 | 2388 | 160 | |
GenBank mRNA | AF102177.1 | - | 18 - 19 | 2565 | 160 |
AK002205.1 | - | 7 - 8 | 813 | 160 | |
AK025545.1 | - | 13 - 14 | 1600 | 160 | |
AK094019.1 | - | 4 - 5 | 1119 | 160 | |
AK124436.1 | - | 8 - 9 | 3856 | 160 | |
AK292669.1 | - | 20 - 21 | 2716 | 160 | |
AK292889.1 | - | 20 - 21 | 2767 | 160 | |
AL359939.1 | - | 6 - 7 | 530 | 160 | |
AY444798.1 | - | 18 - 19 | 2565 | 160 | |
BC030275.1 | - | 19 - 20 | 2675 | 160 | |
BC041868.1 | - | 16 - 17 | 2676 | 160 | |
CR749701.1 | - | 6 - 7 | 530 | 160 |