The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005739.1 | NP_001005739.1 | 22 - 23 | 2959 | 157 |
NM_016516.2 | NP_057600.2 | 22 - 23 | 2995 | 157 | |
XM_006712029.3 | XP_006712092.1 | 21 - 22 | 2855 | 157 | |
XM_017004316.1 | XP_016859805.1 | 21 - 22 | 2675 | 157 | |
GenBank mRNA | AK002205.1 | - | 10 - 11 | 1100 | 157 |
AK025545.1 | - | 16 - 17 | 1887 | 157 | |
AK124436.1 | - | 11 - 12 | 4143 | 157 | |
AK292669.1 | - | 23 - 24 | 3003 | 157 | |
AL137604.1 | - | 1 - 2 | 114 | 157 | |
AL359939.1 | - | 9 - 10 | 817 | 157 | |
AY444798.1 | - | 21 - 22 | 2852 | 157 | |
BC030275.1 | - | 22 - 23 | 2962 | 157 | |
BC041868.1 | - | 19 - 20 | 2963 | 157 | |
CR749701.1 | - | 9 - 10 | 817 | 157 |