The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242581.1 | NP_001229510.1 | 2 - 3 | 347 | 63 |
NM_018993.3 | NP_061866.1 | 1 - 2 | 87 | 63 | |
XM_005260731.2 | XP_005260788.1 | 3 - 4 | 543 | 63 | |
XM_006723574.3 | XP_006723637.1 | 4 - 5 | 525 | 63 | |
XM_006723575.3 | XP_006723638.1 | 3 - 4 | 311 | 63 | |
XM_006723577.2 | XP_006723640.1 | 2 - 3 | 160 | 63 | |
XM_011529255.2 | XP_011527557.2 | 2 - 3 | 620 | 63 | |
XM_011529257.1 | XP_011527559.1 | 2 - 3 | 216 | 63 | |
XM_011529258.2 | XP_011527560.1 | 2 - 3 | 228 | 63 | |
XM_017027887.1 | XP_016883376.1 | 5 - 6 | 866 | 63 | |
XM_017027888.1 | XP_016883377.1 | 6 - 7 | 938 | 63 | |
XM_017027890.1 | XP_016883379.1 | 3 - 4 | 298 | 63 | |
XM_017027891.1 | XP_016883380.1 | 2 - 3 | 167 | 63 | |
XM_017027893.1 | XP_016883382.1 | 2 - 3 | 645 | 63 | |
GenBank mRNA | AB060339.1 | - | 1 - 2 | 51 | 63 |
AK094884.1 | - | 2 - 3 | 348 | 63 | |
BC128065.1 | - | 1 - 2 | 87 | 63 |