The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000016.5 | NP_000007.1 | 8 - 9 | 1144 | 98 |
NM_001127328.2 | NP_001120800.1 | 8 - 9 | 1156 | 98 | |
NM_001286042.1 | NP_001272971.1 | 7 - 8 | 1056 | 98 | |
NM_001286043.1 | NP_001272972.1 | 9 - 10 | 1243 | 98 | |
NM_001286044.1 | NP_001272973.1 | 5 - 6 | 874 | 98 | |
GenBank mRNA | AF251043.1 | - | 8 - 9 | 723 | 98 |
AK296045.1 | - | 5 - 6 | 454 | 98 | |
AK301005.1 | - | 5 - 6 | 612 | 98 | |
AK301039.1 | - | 8 - 9 | 800 | 98 | |
AK301717.1 | - | 7 - 8 | 648 | 98 | |
AK311022.1 | - | 7 - 8 | 626 | 98 | |
AK312629.1 | - | 8 - 9 | 734 | 98 | |
AK315946.1 | - | 7 - 8 | 697 | 98 | |
BC005377.1 | - | 8 - 9 | 738 | 98 | |
BX647734.1 | - | 9 - 10 | 882 | 98 | |
KU177876.1 | - | 8 - 9 | 703 | 98 | |
KU177877.1 | - | 7 - 8 | 615 | 98 | |
KU177878.1 | - | 4 - 5 | 346 | 98 | |
M16827.1 | - | 8 - 9 | 721 | 98 |