Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HES6
Entrez Gene ID
55502
Gene Name
hes family bHLH transcription factor 6
Gene Aliases
C-HAIRY1, HES-6, bHLHb41, bHLHc23
Chromosome Location
Chr.2: 238238267 - 238240124 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142853.2 NP_001136325.1 3 - 4 465 138
NM_018645.5 NP_061115.2 3 - 4 471 138
GenBank mRNA AB035179.1 - 3 - 4 252 138
AF260237.1 - 3 - 4 312 138
AK293111.1 - 3 - 4 385 138
BC007939.2 - 3 - 4 376 138
BE796220.1 - 3 - 4 471 138
DT217800.1 - 3 - 4 388 138

View Full Product Details