Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACC1
Entrez Gene ID
6867
Gene Name
transforming acidic coiled-coil containing protein 1
Gene Aliases
Ga55
Chromosome Location
Chr.8: 38728186 - 38853028 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122824.1 NP_001116296.1 10 - 11 1492 85
NM_001146216.2 NP_001139688.1 12 - 13 2668 85
NM_006283.2 NP_006274.2 12 - 13 2722 85
XM_005273625.4 XP_005273682.1 14 - 15 2872 85
XM_005273628.1 XP_005273685.1 13 - 14 2379 85
XM_005273629.1 XP_005273686.1 14 - 15 2392 85
XM_005273630.1 XP_005273687.1 13 - 14 2400 85
XM_011544631.1 XP_011542933.1 13 - 14 2758 85
XM_011544632.2 XP_011542934.1 15 - 16 2933 85
XM_011544633.1 XP_011542935.1 13 - 14 2671 85
XM_011544634.1 XP_011542936.1 12 - 13 2635 85
XM_011544635.1 XP_011542937.1 13 - 14 2462 85
XM_011544636.2 XP_011542938.1 10 - 11 1118 85
XM_011544637.1 XP_011542939.1 10 - 11 1405 85
XM_011544638.2 XP_011542940.1 10 - 11 1008 85
XM_017013766.1 XP_016869255.1 12 - 13 2343 85
XM_017013767.1 XP_016869256.1 13 - 14 2356 85
XM_017013768.1 XP_016869257.1 12 - 13 2256 85
XM_017013769.1 XP_016869258.1 13 - 14 2269 85
XM_017013770.1 XP_016869259.1 12 - 13 2292 85
XM_017013771.1 XP_016869260.1 12 - 13 2564 85
XM_017013772.1 XP_016869261.1 11 - 12 2256 85
XM_017013773.1 XP_016869262.1 11 - 12 2523 85
XM_017013774.1 XP_016869263.1 11 - 12 2208 85
XM_017013775.1 XP_016869264.1 12 - 13 2277 85
XM_017013776.1 XP_016869265.1 13 - 14 2269 85
XM_017013777.1 XP_016869266.1 11 - 12 2433 85
XM_017013778.1 XP_016869267.1 11 - 12 2132 85
GenBank mRNA AB029026.1 - 11 - 12 1289 85
AF049910.1 - 12 - 13 2663 85
AK294931.1 - 12 - 13 2236 85
AK295071.1 - 12 - 13 2270 85
AK304725.1 - 9 - 10 977 85
AK304774.1 - 10 - 11 1042 85
AK314620.1 - 12 - 13 2722 85
AY039239.1 - 11 - 12 1032 85
AY139007.1 - 4 - 5 1367 85
AY177411.1 - 10 - 11 1433 85
BC041391.1 - 14 - 15 2832 85
BX647696.1 - 12 - 13 2667 85

View Full Product Details