The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122824.1 | NP_001116296.1 | 3 - 4 | 812 | 120 |
NM_001146216.2 | NP_001139688.1 | 5 - 6 | 1988 | 120 | |
NM_006283.2 | NP_006274.2 | 5 - 6 | 2042 | 120 | |
XM_005273625.4 | XP_005273682.1 | 7 - 8 | 2192 | 120 | |
XM_005273628.1 | XP_005273685.1 | 6 - 7 | 1699 | 120 | |
XM_005273629.1 | XP_005273686.1 | 7 - 8 | 1712 | 120 | |
XM_005273630.1 | XP_005273687.1 | 6 - 7 | 1720 | 120 | |
XM_011544631.1 | XP_011542933.1 | 6 - 7 | 2078 | 120 | |
XM_011544632.2 | XP_011542934.1 | 8 - 9 | 2253 | 120 | |
XM_011544635.1 | XP_011542937.1 | 6 - 7 | 1782 | 120 | |
XM_011544636.2 | XP_011542938.1 | 3 - 4 | 438 | 120 | |
XM_017013766.1 | XP_016869255.1 | 5 - 6 | 1663 | 120 | |
XM_017013767.1 | XP_016869256.1 | 6 - 7 | 1676 | 120 | |
XM_017013770.1 | XP_016869259.1 | 5 - 6 | 1612 | 120 | |
XM_017013771.1 | XP_016869260.1 | 5 - 6 | 1884 | 120 | |
XM_017013772.1 | XP_016869261.1 | 4 - 5 | 1576 | 120 | |
XM_017013773.1 | XP_016869262.1 | 4 - 5 | 1843 | 120 | |
XM_017013774.1 | XP_016869263.1 | 4 - 5 | 1528 | 120 | |
GenBank mRNA | AB029026.1 | - | 4 - 5 | 612 | 120 |
AF049910.1 | - | 5 - 6 | 1983 | 120 | |
AK295071.1 | - | 5 - 6 | 1590 | 120 | |
AK303596.1 | - | 4 - 5 | 1474 | 120 | |
AK304725.1 | - | 2 - 3 | 300 | 120 | |
AK304774.1 | - | 3 - 4 | 362 | 120 | |
AK314620.1 | - | 5 - 6 | 2042 | 120 | |
AY039239.1 | - | 4 - 5 | 352 | 120 | |
AY072873.2 | - | 3 - 4 | 454 | 120 | |
AY072874.1 | - | 3 - 4 | 443 | 120 | |
AY072875.1 | - | 4 - 5 | 441 | 120 | |
AY177411.1 | - | 3 - 4 | 753 | 120 | |
BX647696.1 | - | 5 - 6 | 1987 | 120 |