The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318369.1 | NP_001305298.1 | 11 - 12 | 2000 | 78 |
NM_004211.4 | NP_004202.3 | 12 - 13 | 2139 | 78 | |
XM_017018544.1 | XP_016874033.1 | 8 - 9 | 1070 | 78 | |
XM_017018545.1 | XP_016874034.1 | 8 - 9 | 1063 | 78 | |
GenBank mRNA | AF085412.1 | - | 12 - 13 | 2147 | 78 |
AF117999.1 | - | 11 - 12 | 1866 | 78 | |
AF142501.1 | - | 11 - 12 | 1884 | 78 | |
AF352733.1 | - | 12 - 13 | 1866 | 78 | |
BC096319.3 | - | 12 - 13 | 1945 | 78 | |
BC096320.3 | - | 12 - 13 | 1945 | 78 | |
BC096321.1 | - | 12 - 13 | 1945 | 78 |