Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GMEB2
Entrez Gene ID
26205
Gene Name
glucocorticoid modulatory element binding protein 2
Gene Aliases
GMEB-2, P79PIF, PIF79
Chromosome Location
Chr.20: 63587594 - 63627101 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012384.4 NP_036516.1 8 - 9 1037 126
XM_005260202.4 XP_005260259.1 8 - 9 1248 126
XM_006723776.2 XP_006723839.1 6 - 7 689 126
XM_011528779.2 XP_011527081.1 6 - 7 753 126
GenBank mRNA AB033095.1 - 8 - 9 914 126
AF173867.1 - 7 - 8 1313 126
AL133646.1 - 6 - 7 761 126
BC036305.1 - 8 - 9 964 126
BQ954281.1 - 7 - 8 739 126

View Full Product Details