The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172309.1 | NP_001165780.1 | 3 - 4 | 403 | 134 |
NM_144573.3 | NP_653174.3 | 4 - 5 | 595 | 134 | |
XM_005271322.3 | XP_005271379.1 | 4 - 5 | 486 | 134 | |
XM_005271323.3 | XP_005271380.1 | 4 - 5 | 486 | 134 | |
XM_005271324.4 | XP_005271381.1 | 3 - 4 | 390 | 134 | |
XM_005271325.3 | XP_005271382.1 | 4 - 5 | 486 | 134 | |
XM_005271326.3 | XP_005271383.1 | 3 - 4 | 321 | 134 | |
XM_005271327.3 | XP_005271384.1 | 4 - 5 | 486 | 134 | |
GenBank mRNA | AF176780.1 | - | 3 - 4 | 274 | 134 |
AK298565.1 | - | 3 - 4 | 372 | 134 | |
AK300728.1 | - | 4 - 5 | 528 | 134 | |
AK307487.1 | - | 3 - 4 | 321 | 134 | |
AY211912.1 | - | 4 - 5 | 448 | 134 | |
BC055084.1 | - | 4 - 5 | 372 | 134 | |
BX648546.1 | - | 4 - 5 | 483 | 134 | |
CR936630.1 | - | 4 - 5 | 505 | 134 | |
DC412178.1 | - | 3 - 4 | 404 | 134 | |
DQ464902.1 | - | 4 - 5 | 482 | 134 |