The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015156.3 | NP_055971.2 | 11 - 12 | 1646 | 93 |
GenBank mRNA | AK026612.1 | - | 1 - 2 | 186 | 93 |
D31888.1 | - | 11 - 12 | 1151 | 93 |