The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015156.3 | NP_055971.2 | 5 - 6 | 890 | 135 |
GenBank mRNA | AF155595.1 | - | 5 - 6 | 653 | 135 |
BC051003.1 | - | 5 - 6 | 888 | 135 | |
BC064495.1 | - | 5 - 6 | 888 | 135 | |
D31888.1 | - | 5 - 6 | 395 | 135 |