The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030943.3 | NP_112205.2 | 6 - 7 | 684 | 113 |
XM_011537202.2 | XP_011535504.1 | 6 - 7 | 929 | 113 | |
XM_011537203.2 | XP_011535505.1 | 6 - 7 | 1185 | 113 | |
GenBank mRNA | AF328788.1 | - | 6 - 7 | 673 | 113 |
AK055802.1 | - | 5 - 6 | 1397 | 113 | |
AY358468.1 | - | 6 - 7 | 671 | 113 | |
BC029948.1 | - | 6 - 7 | 684 | 113 |