Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COQ10A
Entrez Gene ID
93058
Gene Name
coenzyme Q10A
Gene Aliases
-
Chromosome Location
Chr.12: 56266858 - 56270966 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099337.1 NP_001092807.1 1 - 2 150 108
GenBank mRNA BC026922.1 - 1 - 2 150 108

View Full Product Details