Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGBL1
Entrez Gene ID
123624
Gene Name
ATP/GTP binding protein like 1
Gene Aliases
CCP4, FECD8
Chromosome Location
Chr.15: 86079635 - 87031476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011521226.2 XP_011519528.1 14 - 15 2324 84
XM_011521227.2 XP_011519529.1 14 - 15 2324 84
XM_011521228.2 XP_011519530.1 14 - 15 2324 84
XM_017021918.1 XP_016877407.1 14 - 15 2291 84
XM_017021919.1 XP_016877408.1 13 - 14 2240 84
XM_017021920.1 XP_016877409.1 14 - 15 2324 84
XM_017021921.1 XP_016877410.1 14 - 15 2324 84
GenBank mRNA BC128152.1 - 5 - 6 1170 84

View Full Product Details