The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308465.1 | NP_001295394.1 | 6 - 6 | 3187 | 99 |
NM_001308467.1 | NP_001295396.1 | 6 - 6 | 3164 | 99 | |
NM_001308468.1 | NP_001295397.1 | 4 - 4 | 2985 | 99 | |
NM_001308469.1 | NP_001295398.1 | 4 - 4 | 2962 | 99 | |
NM_001308470.1 | NP_001295399.1 | 3 - 3 | 2895 | 99 | |
NM_001308471.1 | NP_001295400.1 | 2 - 2 | 2794 | 99 | |
NM_001308476.1 | NP_001295405.1 | 5 - 5 | 3058 | 99 | |
NM_020377.3 | NP_065110.1 | 5 - 5 | 3035 | 99 | |
GenBank mRNA | AB038269.1 | - | 1 - 1 | 2748 | 99 |
BC069160.1 | - | 1 - 1 | 2489 | 99 |